Assay Details
Target Gene Details
Entrez Gene ID: | 320095 |
Gene Name: | RIKEN cDNA 6430550D23 gene |
Gene Aliases: |
- |
Location: |
Chr.2:155982970-156007977 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 6430550D23Rik |
Target Gene Details
Entrez Gene ID: | 16328 |
Gene Name: | centrosomal protein 250 |
Gene Aliases: |
AW490617, B230210E21Rik, Cep2, Inmp |
Location: |
Chr.2:155956272-155998900 on Build GRCm38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cep250 | NM_001129999.1 | 24 | 3665 | NP_001123471.1 |
| NM_001130000.1 | 24 | 3663 | NP_001123472.1 | |
| NM_008383.3 | 24 | 3605 | NP_032409.3 | |
| NM_177217.3 | 24 | 3600 | NP_796191.2 | |
| AK161443.1 | 23 | 4248 | ||
| BC150746.1 | 23 | 3427 | ||
| DQ148475.1 | 23 | 3400 | ABA29340.1 |
More Information
Set Membership: |
|

Genomic Map