Assay Details
Target Gene Details
Entrez Gene ID: | 16328 |
Gene Name: | centrosomal protein 250 |
Gene Aliases: |
AW490617, B230210E21Rik, Cep2, Inmp |
Location: |
Chr.2:155956272-155998900 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cep250 | NM_001129999.1 | 7 | 974 | NP_001123471.1 |
| NM_001130000.1 | 7 | 969 | NP_001123472.1 | |
| NM_008383.3 | 7 | 974 | NP_032409.3 | |
| NM_177217.3 | 7 | 969 | NP_796191.2 | |
| AK033281.1 | 7 | 938 | BAC28218.1 | |
| AK045557.1 | 7 | 961 | BAC32417.1 | |
| AK145562.1 | 7 | 970 | ||
| AK161443.1 | 7 | 1075 | ||
| BC150746.1 | 6 | 796 | ||
| DQ148475.1 | 6 | 769 | ABA29340.1 |
More Information
Set Membership: |
|

Genomic Map