Assay Details
Target Gene Details
Entrez Gene ID: | 23856 |
Gene Name: | death inducer-obliterator 1 |
Gene Aliases: |
6720461J16Rik, D130048F08Rik, DATF-1, DIO-1, Datf1, dido |
Location: |
Chr.2:180657963-180710035 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dido1 | NM_175551.4 | 16 | 5396 | NP_780760.2 |
| AK031764.1 | 1 | 434 | ||
| AK032843.1 | 1 | 619 | BAC28053.1 | |
| AY425952.1 | 16 | 5298 | AAR84050.1 |
More Information
Set Membership: |
|

Genomic Map