Assay Details
Target Gene Details
Entrez Gene ID: | 241520 |
Gene Name: | family with sequence similarity 171, member B |
Gene Aliases: |
D430039N05Rik |
Location: |
Chr.2:83812728-83883486 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam171b | NM_175514.2 | 8 | 1892 | NP_780723.1 |
| AK034250.1 | 1 | 165 | BAC28648.1 | |
| AK052508.1 | 8 | 1893 | BAC35021.1 | |
| BC038086.1 | 1 | 582 | AAH38086.1 | |
| BC113787.1 | 8 | 1905 |

Genomic Map