Assay Details
Target Gene Details
Entrez Gene ID: | 241303 |
Gene Name: | family with sequence similarity 78, member A |
Gene Aliases: |
A130092J06Rik |
Location: |
Chr.2:32052166-32087436 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam78a | NM_175511.4 | 2 | 3526 | NP_780720.1 |
| AK077626.1 | 2 | 3531 | ||
| AK079514.1 | 2 | 3527 | BAC37667.1 |

Genomic Map