Assay Details
Target Gene Details
Entrez Gene ID: | 72630 |
Gene Name: | heat shock protein 12B |
Gene Aliases: |
2700081N06Rik |
Location: |
Chr.2:131127412-131145985 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hspa12b | NM_028306.3 | 11 | 1374 | NP_082582.1 |
| AK012548.1 | 11 | 1347 | BAB28308.1 | |
| AY196790.1 | 10 | 1254 | AAO37639.1 | |
| BC011103.1 | 11 | 1374 | AAH11103.1 |

Genomic Map