Assay Details
Target Gene Details
Entrez Gene ID: | 117004 |
Gene Name: | olfactory receptor 73 |
Gene Aliases: |
MOR174-9, fascin-1, mOR-EG |
Location: |
Chr.2:88034196-88037686 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Olfr73 | NM_054090.1 | 1 | 634 | NP_473431.1 |
| AB061228.1 | 1 | 634 | BAB59037.1 |
More Information
Set Membership: |
|

Genomic Map