Assay Details
Target Gene Details
Entrez Gene ID: | 69327 |
Gene Name: | RIKEN cDNA 1700007K13 gene |
Gene Aliases: |
Rbest47 |
Location: |
Chr.2:28462001-28466324 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1700007K13Rik | NM_027040.1 | 1 | 46 | NP_081316.1 |
| AK005731.1 | 1 | 57 | BAB24210.1 | |
| AY927869.1 | AAX22218.1 | |||
| BC099566.1 | 1 | 46 | AAH99566.1 |
Target Gene Details
Entrez Gene ID: | 118451 |
Gene Name: | mitochondrial ribosomal protein S2 |
Gene Aliases: |
1500019M10Rik |
Location: |
Chr.2:28465956-28471177 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrps2 | AK077219.1 | BAC36691.1 |
More Information
Set Membership: |
|

Genomic Map