Assay Details
Target Gene Details
Entrez Gene ID: | 11477 |
Gene Name: | activin A receptor, type 1 |
Gene Aliases: |
ALK2, ActR-I, ActRIA, Acvr, Acvrlk2, Alk-2, Alk8, D330013D15Rik, SKR1, Tsk7L |
Location: |
Chr.2:58446438-58566828 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acvr1 | NM_001110204.1 | 10 | 1503 | NP_001103674.1 |
| NM_001110205.1 | 8 | 981 | NP_001103675.1 | |
| NM_007394.3 | 9 | 1258 | NP_031420.2 | |
| AF332087.1 | 7 | 891 | AAK56115.1 | |
| AF332088.1 | 7 | 891 | AAK56116.1 | |
| AK053428.1 | 10 | 1502 | ||
| AK150014.1 | 9 | 1258 | BAE29239.1 | |
| AK150075.1 | 9 | 1258 | BAE29286.1 | |
| BC058718.1 | 7 | 979 | AAH58718.1 | |
| L15436.1 | 8 | 1331 | AAA40495.1 |
More Information
Set Membership: |
|

Genomic Map