Assay Details
Target Gene Details
Entrez Gene ID: | 20536 |
Gene Name: | solute carrier family 4 (anion exchanger), member 3 |
Gene Aliases: |
A930038D23Rik, Ae3 |
Location: |
Chr.1:75546041-75559431 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc4a3 | NM_009208.3 | 4 | 566 | NP_033234.2 |
| AK045583.1 | 3 | 453 | BAC32423.1 | |
| BC080271.1 | 4 | 561 | AAH80271.1 | |
| M28383.1 | 4 | 536 | AAA37184.1 |
More Information
Set Membership: |
|

Genomic Map