Assay Details
Target Gene Details
Entrez Gene ID: | 110895 |
Gene Name: | solute carrier family 9 (sodium/hydrogen exchanger), member 4 |
Gene Aliases: |
AW990558, D730009J23Rik, NHE4 |
Location: |
Chr.1:40579715-40630731 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc9a4 | NM_177084.3 | 1 | 175 | NP_796058.1 |
| AK085681.1 | 1 | 201 | BAC39504.1 | |
| AK135750.1 | 1 | 201 | ||
| AK141534.1 | 1 | 176 | BAE24721.1 | |
| BC120543.1 | 1 | 93 |
More Information
Set Membership: |
|

Genomic Map