Assay Details
Target Gene Details
Entrez Gene ID: | 170706 |
Gene Name: | transmembrane protein 37 |
Gene Aliases: |
AI173373, AW540203, Pr, Pr1 |
Location: |
Chr.1:120067377-120074069 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem37 | NM_019432.2 | 2 | 386 | NP_062305.1 |
| AJ272046.1 | 2 | 338 | CAB86387.1 | |
| AK002424.1 | 2 | 405 | BAB22091.1 | |
| AK050134.1 | 2 | 386 | BAC34086.1 | |
| AK086093.1 | 2 | 391 | BAC39608.1 | |
| BC024613.1 | 2 | 382 | AAH24613.1 |
More Information
Set Membership: |
|

Genomic Map