Assay Details
Target Gene Details
Entrez Gene ID: | 108699 |
Gene Name: | chimerin 1 |
Gene Aliases: |
0610007I19Rik, 0710001E19Rik, 1700112L09Rik, 2900046J01Rik, AI413815, ARHGAP2 |
Location: |
Chr.2:73610660-73775367 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chn1 | NM_001113246.2 | 13 | 2743 | NP_001106717.2 |
| NM_001166603.1 | 12 | 2569 | NP_001160075.1 | |
| NM_001166604.1 | 6 | 2381 | NP_001160076.1 | |
| NM_029716.3 | 6 | 1944 | NP_083992.1 | |
| NM_175752.3 | 7 | 2555 | NP_786928.2 | |
| AK032630.1 | 7 | 2513 | ||
| AK049943.1 | 7 | 2499 | BAC33996.1 |
Target Gene Details
Entrez Gene ID: | 329427 |
Gene Name: | chimerin 1, opposite strand 3 |
Gene Aliases: |
Gm13704, OTTMUSG00000013429 |
Location: |
Chr.2:73596526-73616012 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chn1os3 | NR_040623.1 |
More Information
Set Membership: |
|

Genomic Map