Assay Details
Target Gene Details
Entrez Gene ID: | 12672 |
Gene Name: | cholinergic receptor, muscarinic 4 |
Gene Aliases: |
Chrm-4, M4 |
Location: |
Chr.2:91919619-91929835 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chrm4 | NM_007699.2 | 2 | 590 | NP_031725.1 |
| BC120504.1 | 1 | 434 | ||
| BC137677.1 | 1 | 434 |
More Information
Set Membership: |
|

Genomic Map