Assay Details
Target Gene Details
Entrez Gene ID: | 11435 |
Gene Name: | cholinergic receptor, nicotinic, alpha polypeptide 1 (muscle) |
Gene Aliases: |
AI385656, AI608266, Achr-1, Acra |
Location: |
Chr.2:73563281-73580338 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chrna1 | NM_007389.5 | 9 | 2314 | NP_031415.2 |
| AK029177.1 | 9 | 2283 | BAC26337.1 | |
| BC125439.1 | 9 | 2314 | ||
| BC137678.1 | 9 | 2314 |
More Information
Set Membership: |
|

Genomic Map