Assay Details
Target Gene Details
Entrez Gene ID: | 23856 |
Gene Name: | death inducer-obliterator 1 |
Gene Aliases: |
6720461J16Rik, D130048F08Rik, DATF-1, DIO-1, Datf1, dido |
Location: |
Chr.2:180657963-180710035 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dido1 | NM_001291433.1 | 15 | 4743 | NP_001278362.1 |
| NM_175551.4 | NP_780760.2 | |||
| NM_177852.4 | 15 | 4691 | NP_808520.2 | |
| NR_111965.1 | 13 | 4441 | ||
| AK049707.1 | 9 | 3358 | ||
| AK129117.1 | 15 | 4691 | BAC97927.1 | |
| AY425952.1 | AAR84050.1 | |||
| BC044755.1 | 9 | 2842 | AAH44755.1 |
More Information
Set Membership: |
|

Genomic Map