Assay Details
Target Gene Details
Entrez Gene ID: | 23856 |
Gene Name: | death inducer-obliterator 1 |
Gene Aliases: |
6720461J16Rik, D130048F08Rik, DATF-1, DIO-1, Datf1, dido |
Location: |
Chr.2:180657963-180710035 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dido1 | NM_001291432.1 | 4 | 1165 | NP_001278361.1 |
| NM_001291433.1 | 4 | 1217 | NP_001278362.1 | |
| NM_011805.3 | 4 | 1217 | NP_035935.2 | |
| NM_175551.4 | 4 | 1217 | NP_780760.2 | |
| NM_177852.4 | 4 | 1165 | NP_808520.2 | |
| NR_111965.1 | 3 | 1075 | ||
| AJ238332.1 | 4 | 1119 | CAB48401.1 | |
| AK014422.1 | 4 | 1215 | ||
| AK042474.1 | 4 | 1186 | BAC31270.1 | |
| AK129117.1 | 4 | 1165 | BAC97927.1 | |
| AY425951.1 | 4 | 1119 | AAR84049.1 | |
| AY425952.1 | 4 | 1119 | AAR84050.1 | |
| BC029110.1 | 5 | 1219 | ||
| BC096662.1 | 4 | 1157 | AAH96662.1 | |
| BC138712.1 | 3 | 994 | ||
| BC138713.1 | 3 | 994 |
More Information
Set Membership: |
|

Genomic Map