Assay Details
Target Gene Details
Entrez Gene ID: | 69408 |
Gene Name: | DnaJ (Hsp40) homolog, subfamily C, member 17 |
Gene Aliases: |
1700025B16Rik, C87112, D9Bwg1371e |
Location: |
Chr.2:119172500-119208795 on Build GRCm38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dnajc17 | NM_139139.2 | NP_631878.2 | ||
| AK077385.1 | BAC36778.1 | |||
| BC013487.1 | AAH13487.1 | |||
| BP770408.1 |
Target Gene Details
Entrez Gene ID: | 623781 |
Gene Name: | predicted gene 14137 |
Gene Aliases: |
OTTMUSG00000015762 |
Location: |
Chr.2:119174509-119177575 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm14137 | NM_001039223.3 | 1 | 1023 | NP_001034312.1 |
| AK028753.1 | 1 | 1024 | BAC26099.1 | |
| BC150984.1 | 1 | 1013 | ||
| BC150992.1 | 1 | 1013 |
More Information
Set Membership: |
|

Genomic Map