Assay Details
Target Gene Details
Entrez Gene ID: | 100503468 |
Gene Name: | predicted gene 14023 |
Gene Aliases: |
OTTMUSG00000015408 |
Location: |
Chr.2:129297370-129307826 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm14023 | NR_040371.1 | |||
| AK042010.1 |
Target Gene Details
Entrez Gene ID: | 16175 |
Gene Name: | interleukin 1 alpha |
Gene Aliases: |
Il-1a |
Location: |
Chr.2:129299609-129310901 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Il1a | NM_010554.4 | 7 | 899 | NP_034684.2 |
| AK156449.1 | 7 | 900 | BAE33715.1 | |
| BC003727.1 | 6 | 868 | AAH03727.1 | |
| M14639.1 | AAA40058.1 | |||
| X01450.1 | 7 | 899 | CAA25682.1 |
More Information
Set Membership: |
|

Genomic Map