Assay Details
Target Gene Details
Entrez Gene ID: | 16176 |
Gene Name: | interleukin 1 beta |
Gene Aliases: |
IL-1beta, Il-1b |
Location: |
Chr.2:129363843-129371173 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Il1b | NM_008361.3 | 7 | 891 | NP_032387.1 |
| AK156396.1 | 6 | 1443 | ||
| AK157245.1 | 4 | 3313 | ||
| AK224980.1 | 6 | 1443 | ||
| AK225002.1 | 7 | 910 | ||
| AK225003.1 | 3 | 485 | ||
| BC011437.1 | 7 | 891 | AAH11437.1 | |
| M15131.1 | 7 | 900 | AAA39276.1 |
More Information
Set Membership: |
|

Genomic Map