Assay Details
Target Gene Details
Entrez Gene ID: | 17147 |
Gene Name: | melanoma antigen, family B, 3 |
Gene Aliases: |
Mage-b3, Mage-ps1, Mage-rs3, Smage3 |
Location: |
Chr.2:121953771-121956092 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mageb3 | NM_008545.2 | 1 | 774 | NP_032571.2 |
| AF311315.3 | 1 | 42 | AAG60640.2 | |
| AF311316.2 | 1 | 42 | AAG60641.1 | |
| AK019658.1 | 1 | 774 | BAB31825.1 | |
| AY099462.1 | 1 | 42 | AAM47609.1 | |
| AY099463.1 | 1 | 42 | AAM47610.1 | |
| AY099464.1 | 1 | 42 | AAM47611.1 | |
| BC141002.1 | 1 | 672 |
More Information
Set Membership: |
|

Genomic Map