Assay Details
Target Gene Details
Entrez Gene ID: | 227619 |
Gene Name: | mannosidase, alpha, class 1B, member 1 |
Gene Aliases: |
E430019H13Rik, ERMan1, Gm108, MANA-ER |
Location: |
Chr.2:25332743-25352213 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Man1b1 | NM_001029983.2 | 13 | 3121 | NP_001025154.1 |
| AK088523.1 | 8 | 2407 | BAC40402.1 | |
| BC006645.1 | 11 | 2737 | AAH06645.1 | |
| BC025500.1 | 5 | 1773 | AAH25500.1 |
More Information
Set Membership: |
|

Genomic Map