Assay Details
Target Gene Details
Entrez Gene ID: | 20204 |
Gene Name: | paired related homeobox 2 |
Gene Aliases: |
Prx2, S8 |
Location: |
Chr.2:30845064-30881251 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prrx2 | NM_009116.2 | 1 | 213 | NP_033142.2 |
| BC137872.1 | 1 | 240 | ||
| BC137874.1 | 1 | 240 | ||
| CJ179520.1 | 1 | 214 | ||
| X52875.1 | 1 | 299 | CAA37055.1 |

Genomic Map