Assay Details
Target Gene Details
Entrez Gene ID: | 67326 |
Gene Name: | RIKEN cDNA 1700037H04 gene |
Gene Aliases: |
- |
Location: |
Chr.2:131146315-131160086 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1700037H04Rik | NM_026091.2 | 6 | 922 | NP_080367.1 |
| AK003406.1 | 6 | 923 | BAB22770.1 | |
| AK040365.1 | 3 | 1328 | ||
| AK048328.1 | 1 | 196 | ||
| BC006041.1 | 5 | 834 | AAH06041.1 | |
| BC071212.1 | 7 | 1038 | AAH71212.1 |

Genomic Map