Assay Details
Target Gene Details
Entrez Gene ID: | 240873 |
Gene Name: | tumor necrosis factor (ligand) superfamily, member 18 |
Gene Aliases: |
Gitrl |
Location: |
Chr.1:161494657-161505290 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tnfsf18 | NM_183391.3 | 3 | 1133 | NP_899247.3 |
| AJ577579.1 | 3 | 1135 | CAE12166.1 |

Genomic Map