Assay Details
Target Gene Details
Entrez Gene ID: | 11438 |
Gene Name: | cholinergic receptor, nicotinic, alpha polypeptide 4 |
Gene Aliases: |
Acra-4, Acra4, EBN1, ENFL1 |
Location: |
Chr.2:181022311-181043580 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chrna4 | NM_015730.5 | 6 | 2729 | NP_056545.3 |
| AK034228.1 | 6 | 2740 | BAC28638.1 | |
| AK083157.1 | 6 | 2660 | BAC38788.1 | |
| AK136886.1 | 6 | 2716 | BAE23155.1 | |
| AK143938.1 | 6 | 2737 | BAE25619.1 | |
| BC053013.1 | 6 | 2729 | AAH53013.1 |
More Information
Set Membership: |
|

Genomic Map