Assay Details
Target Gene Details
Entrez Gene ID: | 18019 |
Gene Name: | nuclear factor of activated T cells, cytoplasmic, calcineurin dependent 2 |
Gene Aliases: |
AI607462, NF-ATc2, NF-ATp, NFAT1, NFAT1-D, Nfatp |
Location: |
Chr.2:168476410-168601657 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nfatc2 | NM_001136073.2 | 11 | 5549 | NP_001129545.1 |
| NM_001291168.1 | 10 | 5348 | NP_001278097.1 | |
| NM_001291169.1 | 11 | 5434 | NP_001278098.1 | |
| NM_001291170.1 | 10 | 4828 | NP_001278099.1 | |
| NM_001291171.1 | 10 | 4398 | NP_001278100.1 | |
| NM_001291173.1 | 9 | 4312 | NP_001278102.1 | |
| NM_001291174.1 | 11 | 4914 | NP_001278103.1 | |
| NM_001291175.1 | 11 | 5029 | NP_001278104.1 | |
| NM_001291176.1 | 10 | 4513 | NP_001278105.1 | |
| NM_001291177.1 | 10 | 4943 | NP_001278106.1 | |
| NM_001291178.1 | 9 | 4427 | NP_001278107.1 | |
| NM_010899.3 | 10 | 5463 | NP_035029.2 | |
| AK161174.1 | 9 | 4124 | BAE36226.1 | |
| EU887581.1 | 11 | 5315 | ||
| EU887582.1 | 11 | 5375 | ||
| EU887583.1 | 11 | 4912 | ||
| EU887584.1 | 11 | 5012 | ||
| EU887585.1 | 10 | 4396 | ||
| EU887586.1 | 10 | 4496 |
More Information
Set Membership: |
|

Genomic Map