Assay Details
Target Gene Details
Entrez Gene ID: | 18041 |
Gene Name: | nitrogen fixation gene 1 (S. cerevisiae) |
Gene Aliases: |
AA987187, m-Nfs1, m-Nfsl |
Location: |
Chr.2:156123637-156144186 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nfs1 | NM_010911.2 | 13 | 1707 | NP_035041.2 |
| AJ222660.1 | 13 | 1706 | CAA10916.1 | |
| AK004077.1 | 1 | 332 | BAB23157.1 | |
| AK005724.1 | 13 | 1697 | ||
| AK075575.1 | 13 | 1705 | BAC35831.1 | |
| AK132838.1 | 13 | 1708 | BAE21386.1 | |
| AK148527.1 | 13 | 1708 | BAE28601.1 | |
| AK159828.1 | 13 | 1696 | BAE35407.1 | |
| AK181025.1 | 1 | 201 | ||
| AK211463.1 | ||||
| BC010586.1 | 13 | 1685 | AAH10586.1 |
More Information
Set Membership: |
|

Genomic Map