Assay Details
Target Gene Details
Entrez Gene ID: | 329581 |
Gene Name: | baculoviral IAP repeat-containing 7 (livin) |
Gene Aliases: |
E130019N06, KIAP, Livin, ML-IAP |
Location: |
Chr.2:180928152-180935028 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Birc7 |
Target Gene Details
Entrez Gene ID: | 58237 |
Gene Name: | Na+/K+ transporting ATPase interacting 4 |
Gene Aliases: |
AB030182, C030019F02Rik |
Location: |
Chr.2:180934769-180954888 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nkain4 | NM_001141933.1 | 6 | 690 | NP_001135405.1 |
| NM_021426.4 | 7 | 775 | NP_067401.3 | |
| AB030182.1 | 7 | 753 | BAA92746.1 | |
| AK005262.1 | 7 | 746 | BAB23914.1 | |
| AK007632.1 | 7 | 740 | BAB25149.1 | |
| AK021096.1 | 7 | 770 | BAB32298.1 | |
| BC061107.1 | 7 | 760 | AAH61107.1 | |
| BC089330.1 | 7 | 727 | AAH89330.1 |

Genomic Map