Assay Details
Target Gene Details
Entrez Gene ID: | 57329 |
Gene Name: | otoraplin |
Gene Aliases: |
CDRAP, Fdp, MIA, MIAL |
Location: |
Chr.2:143078492-143081700 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Otor | NM_020595.2 | 4 | 458 | NP_065620.1 |
| AF243504.1 | 4 | 458 | AAG42355.1 | |
| AJ243939.1 | 4 | 473 | CAC27444.1 | |
| AK047965.1 | 1 | 2768 | ||
| BC139152.1 | 4 | 557 | ||
| BC139153.1 | 4 | 557 |

Genomic Map