Assay Details
Target Gene Details
Entrez Gene ID: | 64292 |
Gene Name: | prostaglandin E synthase |
Gene Aliases: |
2410099E23Rik, D2Ertd369e, Pges, mPGES, mPGES-1 |
Location: |
Chr.2:30889471-30903297 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptges | NM_022415.3 | 3 | 2527 | NP_071860.1 |
| AK080897.1 | 3 | 2532 | BAC38069.1 | |
| AK167015.1 | 3 | 2396 |
More Information
Set Membership: |
|

Genomic Map