Assay Details
Target Gene Details
Entrez Gene ID: | 56045 |
Gene Name: | SAM domain and HD domain, 1 |
Gene Aliases: |
E330031J07Rik, Mg11 |
Location: |
Chr.2:157097529-157135277 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Samhd1 | NM_001139520.1 | 17 | 3029 | NP_001132992.1 |
| NM_018851.3 | 16 | 2906 | NP_061339.3 | |
| BC067198.1 | 16 | 2829 | AAH67198.1 | |
| BG068904.2 | 1 | 211 |
More Information
Set Membership: |
|

Genomic Map