Assay Details
Target Gene Details
Entrez Gene ID: | 20265 |
Gene Name: | sodium channel, voltage-gated, type I, alpha |
Gene Aliases: |
B230332M13, Nav1.1 |
Location: |
Chr.2:66270781-66440847 on Build GRCm38 |
Assay Gene Location: | Within Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Scn1a | NM_018733.2 | 26 | 4873 | NP_061203.2 |
| AJ810515.1 | 2 | 255 | CAH17959.1 | |
| EF591872.1 | 10 | 1677 | ||
| L42339.1 | AAA67107.1 |
More Information
Set Membership: |
|

Genomic Map