Assay Details
Target Gene Details
Entrez Gene ID: | 11550 |
Gene Name: | adrenergic receptor, alpha 1d |
Gene Aliases: |
Adra-1, Adra1, Adra1a, Gpcr8, Spr8, [a]1d, alpha1D-AR |
Location: |
Chr.2:131545357-131562285 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adra1d | NM_013460.4 | 1 | 603 | NP_038488.4 |
| S80044.1 | 1 | 603 | AAB47042.1 |
More Information
Set Membership: |
|

Genomic Map