Assay Details
Target Gene Details
Entrez Gene ID: | 228061 |
Gene Name: | alkylglycerone phosphate synthase |
Gene Aliases: |
5832437L22, 9930035G10Rik, AW123847, Adaps, Adas, Adhaps, Adps, Aldhpsy, bs2 |
Location: |
Chr.2:75832177-75931350 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Agps | NM_172666.3 | 1 | 228 | NP_766254.2 |
| AK031049.1 | 1 | 230 | BAC27229.1 | |
| AK031154.1 | 1 | 69 | ||
| AK042745.1 | 1 | 68 | ||
| AK053686.1 | 1 | 217 | BAC35474.1 | |
| BC063086.1 | 1 | 213 | AAH63086.1 | |
| BY713473.1 | 1 | 228 |

Genomic Map