Assay Details
Target Gene Details
Entrez Gene ID: | 70852 |
Gene Name: | RIKEN cDNA 4921507L20 gene |
Gene Aliases: |
- |
Location: |
Chr.2:93980034-93983575 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4921507L20Rik | NR_045827.1 | |||
| AK014837.1 | BAB29573.1 |
Target Gene Details
Entrez Gene ID: | 69113 |
Gene Name: | alkB, alkylation repair homolog 3 (E. coli) |
Gene Aliases: |
1700108H04Rik, 1810020C19Rik, Abh3, mABH3 |
Location: |
Chr.2:93980634-94010823 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Alkbh3 | NM_026944.1 | 10 | 948 | NP_081220.1 |
| AK007134.1 | 3 | 255 | ||
| AK007557.1 | 4 | 468 | ||
| BC018196.1 | 10 | 948 | AAH18196.1 |
More Information
Set Membership: |
|

Genomic Map