Assay Details
Target Gene Details
Entrez Gene ID: | 252876 |
Gene Name: | gypsy retrotransposon integrase 1 |
Gene Aliases: |
4930429M06Rik, 8430411H10Rik, Gin-1, Zh2c2 |
Location: |
Chr.1:97770168-97793710 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gin1 | NM_026250.2 | 8 | 1475 | NP_080526.2 |
| AK014287.1 | 8 | 1482 | ||
| AK015243.1 | 8 | 1454 | BAB29761.1 | |
| AK090002.1 | 8 | 1193 | BAE43402.1 | |
| AK169361.1 | 8 | 1438 | BAE41110.1 | |
| BC033572.1 | 7 | 1420 | AAH33572.1 |

Genomic Map