Assay Details
Target Gene Details
Entrez Gene ID: | 58202 |
Gene Name: | negative elongation factor complex member B |
Gene Aliases: |
A730008L03Rik, AB041607, AI663983, Cobra1, Nelf-b |
Location: |
Chr.2:25199712-25211489 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nelfb | NM_021393.2 | 1 | 64 | NP_067368.2 |
| AK046129.1 | ||||
| BU519080.1 | ||||
| BY721962.1 | 1 | 64 |

Genomic Map