Assay Details
Target Gene Details
Entrez Gene ID: | 58202 |
Gene Name: | negative elongation factor complex member B |
Gene Aliases: |
A730008L03Rik, AB041607, AI663983, Cobra1, Nelf-b |
Location: |
Chr.2:25199712-25211489 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nelfb | NM_021393.2 | 13 | 2013 | NP_067368.2 |
| AB041607.1 | 12 | 2134 | BAA95090.1 | |
| AK080419.1 | 13 | 1715 | ||
| AK087524.1 | 13 | 1713 | ||
| AK173124.1 | 12 | 1548 | BAD32402.1 | |
| AK194319.1 | 1 | 129 | ||
| AK206289.1 | 1 | 129 | ||
| AK216002.1 | 1 | 129 | ||
| AK219659.1 | 1 | 129 | ||
| BC004762.1 | 13 | 1901 | AAH04762.1 |

Genomic Map