Assay Details
Target Gene Details
Entrez Gene ID: | 228859 |
Gene Name: | fat storage-inducing transmembrane protein 2 |
Gene Aliases: |
AI414861, D930001I22Rik, Fit2 |
Location: |
Chr.2:163468703-163472629 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fitm2 | NM_173397.4 | 2 | 1168 | NP_775573.1 |
| AK040900.1 | 2 | 1139 | BAC30737.1 | |
| AK043244.1 | 2 | 1169 | BAC31503.1 | |
| AK054016.1 | 2 | 1169 | ||
| AK080142.1 | 2 | 1169 | ||
| AK163611.1 | 2 | 1146 | BAE37420.1 |

Genomic Map