Assay Details
Target Gene Details
Entrez Gene ID: | 228607 |
Gene Name: | mitochondrial antiviral signaling protein |
Gene Aliases: |
D430028G21Rik, IPS-1, Visa, cardif |
Location: |
Chr.2:131234063-131248025 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mavs | NM_001206382.1 | 7 | 906 | NP_001193311.1 |
| NM_001206383.1 | 5 | 640 | NP_001193312.1 | |
| NM_001206385.1 | 6 | 815 | NP_001193314.1 | |
| NM_144888.2 | 6 | 1043 | NP_659137.1 | |
| AK028421.1 | 6 | 814 | BAC25940.1 | |
| AK085038.1 | 6 | 1002 | ||
| AK153748.1 | 7 | 908 | BAE32168.1 | |
| AX775773.1 | 6 | 792 | CAE11586.1 | |
| BC020006.1 | 6 | 775 | AAH20006.1 | |
| BC025825.1 | 6 | 968 | AAH25825.1 | |
| BC031352.1 | 6 | 952 | AAH31352.1 | |
| BC037391.1 | 5 | 754 | AAH37391.1 | |
| DQ167127.1 | 5 | 692 | ABA54891.1 | |
| DQ174271.1 | 5 | 692 | AAZ80418.1 |

Genomic Map