Assay Details
Target Gene Details
Entrez Gene ID: | 109241 |
Gene Name: | methyl-CpG binding domain protein 5 |
Gene Aliases: |
9430004D19Rik, AA536666, AI426407, C030040A15Rik, Gm1630, OTTMUSG00000012483 |
Location: |
Chr.2:48949508-49317070 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mbd5 | NM_001290656.1 | 7 | 3871 | NP_001277585.1 |
| NM_029924.2 | 14 | 5625 | NP_084200.1 | |
| AK173176.1 | 7 | 4492 | BAD32454.1 | |
| BC138375.1 | 7 | 3776 | ||
| BC138378.1 | 7 | 3776 |

Genomic Map