Assay Details
Target Gene Details
Entrez Gene ID: | 18088 |
Gene Name: | NK2 homeobox 2 |
Gene Aliases: |
Nkx-2.2, Nkx2.2, tinman |
Location: |
Chr.2:147177546-147195783 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nkx2-2 | NM_001077632.1 | 3 | 1094 | NP_001071100.1 |
| AK043561.1 | 3 | 948 | BAC31582.1 | |
| AK162904.1 | 2 | 296 | BAE37107.1 | |
| BB183898.1 |
More Information
Set Membership: |
|

Genomic Map