Assay Details
Target Gene Details
Entrez Gene ID: | 18088 |
Gene Name: | NK2 homeobox 2 |
Gene Aliases: |
Nkx-2.2, Nkx2.2, tinman |
Location: |
Chr.2:147177546-147195783 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nkx2-2 | NM_001077632.1 | 1 | 436 | NP_001071100.1 |
| NM_010919.2 | 1 | 436 | NP_035049.1 | |
| AK043561.1 | 1 | 290 | BAC31582.1 | |
| AK141002.1 | 1 | 438 | BAE24543.1 | |
| BC138159.1 | 2 | 257 | ||
| BC138160.1 | 2 | 257 | ||
| U31566.1 | 1 | 393 | AAA79303.1 |
Target Gene Details
Entrez Gene ID: | 100313531 |
Gene Name: | NK2 homeobox 2, opposite strand |
Gene Aliases: |
Gm14112, Nkx2-2as, Nkx2.2AS, OTTMUSG00000015691 |
Location: |
Chr.2:147184083-147331681 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nkx2-2os | NR_030769.2 | |||
| AK045921.1 |
More Information
Set Membership: |
|

Genomic Map