Assay Details
Target Gene Details
Entrez Gene ID: | 18088 |
Gene Name: | NK2 homeobox 2 |
Gene Aliases: |
Nkx-2.2, Nkx2.2, tinman |
Location: |
Chr.2:147177546-147195783 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nkx2-2 | NM_001077632.1 | NP_001071100.1 | ||
| NM_010919.2 | 2 | 1822 | NP_035049.1 | |
| AK043561.1 | BAC31582.1 | |||
| AK141002.1 | 2 | 1827 | BAE24543.1 | |
| AK162904.1 | BAE37107.1 | |||
| BC138159.1 | 3 | 1643 | ||
| BC138160.1 | 3 | 1643 | ||
| U31566.1 | 2 | 1781 | AAA79303.1 |
More Information
Set Membership: |
|

Genomic Map