Assay Details
Target Gene Details
Entrez Gene ID: | 73338 |
Gene Name: | inositol 1,4,5-triphosphate receptor interacting protein-like 1 |
Gene Aliases: |
1700041B20Rik |
Location: |
Chr.2:127138769-127143488 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Itpripl1 | NM_001163527.1 | 2 | 1481 | NP_001156999.1 |
| NM_001163528.1 | 2 | 1305 | NP_001157000.1 | |
| AK006667.1 | 2 | 1306 | ||
| AK028913.1 | 3 | 1221 | ||
| BC130218.1 | 2 | 1330 |

Genomic Map