Assay Details
Target Gene Details
Entrez Gene ID: | 18128 |
Gene Name: | notch 1 |
Gene Aliases: |
9930111A19Rik, Mis6, N1, Tan1, lin-12 |
Location: |
Chr.2:26457902-26503822 on Build GRCm38 |
Assay Gene Location: | Within Exon 31 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Notch1 | NM_008714.3 | 31 | 6115 | NP_032740.3 |
| AB101634.1 | ||||
| AF508809.1 | 31 | 5930 | AAM28905.1 | |
| AK054089.1 | 6 | 1187 | ||
| AK090118.1 | 31 | 6090 | ||
| AK154528.1 | 7 | 1737 | BAE32653.1 | |
| AK157475.1 | 6 | 954 | BAE34095.1 | |
| BC138441.1 | 31 | 5930 | ||
| BC138442.1 | 31 | 5930 | ||
| X82562.1 | CAA57909.1 | |||
| Z11886.1 | 31 | 5929 | CAA77941.1 | |
| Z21925.1 | 2 | 430 |

Genomic Map