Assay Details
Target Gene Details
Entrez Gene ID: | 18146 |
Gene Name: | neural proliferation, differentiation and control 1 |
Gene Aliases: |
AI314472, NPDC-1 |
Location: |
Chr.2:25403050-25409494 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Npdc1 | NM_008721.4 | 1 | 123 | NP_032747.2 |
| AK146466.1 | BAE27193.1 | |||
| BB870287.1 | 1 | 103 | ||
| BC003320.1 | 1 | 90 | AAH03320.1 | |
| CJ057319.1 | 1 | 125 |

Genomic Map