Assay Details
Target Gene Details
Entrez Gene ID: | 74450 |
Gene Name: | pantothenate kinase 2 |
Gene Aliases: |
4933409I19Rik, AI642621 |
Location: |
Chr.2:131262011-131299188 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pank2 | NM_153501.2 | 1 | 212 | NP_705721.2 |
| AK082821.1 | 1 | 218 | ||
| AK154023.1 | 1 | 144 | ||
| AK154080.1 | 1 | 176 | BAE32361.1 | |
| AK159175.1 | 1 | 203 | ||
| AK167861.1 | BAE39879.1 | |||
| BC106184.1 | 1 | 166 | AAI06185.1 | |
| BY728847.1 | 1 | 212 |
More Information
Set Membership: |
|

Genomic Map