Assay Details
Target Gene Details
Entrez Gene ID: | 74450 |
Gene Name: | pantothenate kinase 2 |
Gene Aliases: |
4933409I19Rik, AI642621 |
Location: |
Chr.2:131262011-131299188 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pank2 | NM_153501.2 | 7 | 3282 | NP_705721.2 |
| AK154023.1 | 5 | 2640 | ||
| AK164447.1 | 1 | 532 |
More Information
Set Membership: |
|

Genomic Map